4-Amino-2,5-difluorobenzonitrile ≥98.0% (por GC)
Proveedor: TCI
|
Peligro
|
A2106-1G
A2106-5G
TCIAA2106-1GEA
25
EUR
TCIAA2106-1G
TCIAA2106-5G
4-Amino-2,5-difluorobenzonitrile ≥98.0% (por GC)
4-Amino-2,5-difluorobenzonitrile
Formula:
C₇H₄F₂N₂ MW: 154,12 g/mol |
Número MDL:
MFCD00190102 Núm. CAS: 112279-61-5 UN: 3439 ADR: 6.1,III |
Resultados test especificaciones
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 95.0 to 99.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...