2,6-Difluoro-3-pyridineboronic acid (contains varying amounts of Anhydride)

Proveedor: TCI
Atención

Sinónimos: 2,6-Difluoropyridine-3-boronic acid

D4006-1G D4006-5G
TCIAD4006-1GEA 25 EUR
TCIAD4006-1G TCIAD4006-5G
2,6-Difluoro-3-pyridineboronic acid (contains varying amounts of Anhydride)
2,6-Difluoro-3-pyridineboronic acid
Formula: C₅H₄BF₂NO₂
MW: 158,90 g/mol
Número MDL: MFCD03788245
Núm. CAS: 136466-94-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f344a16cd747784921b97f15f90a969980aee47&vid=0f344a16cd747784921b97f15f90a969980aee47&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) 97.0 to 113.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR