5-Bromo-o-toluic acid ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

B3050-25G B3050-5G
TCIAB3050-25GEA 55 EUR
TCIAB3050-25G TCIAB3050-5G
5-Bromo-o-toluic acid ≥98.0% (by GC, titration analysis)
5-Bromo-o-toluic acid
Formula: C₈H₇BrO₂
Número MDL: MFCD00267350
Núm. CAS: 79669-49-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b044f9a68ac2703a1ec63af822162be143f4b10d&vid=b044f9a68ac2703a1ec63af822162be143f4b10d&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 169.0 to 173.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR