4'-tert-Butylbiphenyl-4-carboxylic acid ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
4'-(tert-Butyl)-[1,1'-biphenyl]-4-carboxylic acid
B1986-1G
B1986-5G
TCIAB1986-1GEA
105
EUR
TCIAB1986-1G
TCIAB1986-5G
4'-tert-Butylbiphenyl-4-carboxylic acid ≥98.0% (by GC, titration analysis)
4'-tert-Butylbiphenyl-4-carboxylic acid
Formula:
C₁₇H₁₈O₂ MW: 254,3200 g/mol |
Número MDL:
MFCD01631300 Núm. CAS: 5748-42-5 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in THF | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...