Methyl-3-amino-4-chlorobenzoate ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
Methyl 3-amino-4-chlorobenzoate, Methyl 3-Amino-4-chlorobenzoate, 3-amino-4-chlorobenzoic acid methylester
M1778-25G
M1778-5G
TCIAM1778-25GEA
160
EUR
TCIAM1778-25G
TCIAM1778-5G
Methyl-3-amino-4-chlorobenzoate ≥98.0% (by GC, titration analysis)
Methyl 3-amino-4-chlorobenzoate
Formula:
C₈H₈ClNO₂ MW: 185,6100 g/mol |
Número MDL:
MFCD03470533 Núm. CAS: 40872-87-5 |
Resultados test especificaciones
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 81.0 to 85.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...