Calix[4]areno ≥98.0% (by HPLC) (contains ca. 8% Chloroform)

Proveedor: TCI
Peligro

Sinónimos: 1,3,5,7(1,3)-Tetrabenzenacyclooctaphane-12,32,52,72-tetraol

C1455-1G C1455-100MG
TCIAC1455-1GEA 101 EUR
TCIAC1455-1G TCIAC1455-100MG
Calix[4]areno ≥98.0% (by HPLC) (contains ca. 8% Chloroform)
Calix[4]areno
Formula: C₂₈H₂₄O₄
MW: 424,5 g/mol
Número MDL: MFCD00233673
Núm. CAS: 74568-07-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd9fdd6d1b39fe90bd4725fd2e7ce807c9cab819&vid=cd9fdd6d1b39fe90bd4725fd2e7ce807c9cab819&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Residual Chloroform max. 10.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR