HPLC columns, BioBasic™ SEC
Proveedor: Thermo Scientific
HYPE73505-154630EA 756 EUR
HYPE73505-154630 HYPE73405-304630 HYPE73505-152130 HYPE73305-152130 HYPE73405-037821 HYPE73405-154630 HYPE73505-254630 HYPE73305-037821 HYPE73305-154630 HYPE73505-304630
HPLC columns, BioBasic™ SEC
BioBasic™ SEC columns are ideal for high efficiency gel filtration separation of proteins, peptides and other biological water soluble molecules.
  • Simple mechanism of interaction based on molecular size and shape
  • Ideal for sample clean-up