Tetraethylammonium toluene-4-sulphonate ≥98.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
Tetraethylammonium p-toluenesulphonate, Tetraethylammonium p-Toluenesulphonate
T0937-25G
T0937-500G
TCIAT0937-25GEA
29
EUR
TCIAT0937-25G
TCIAT0937-500G
Tetraethylammonium toluene-4-sulphonate ≥98.0% (by titrimetric analysis)
Tetraethylammonium toluene-4-sulphonate
Formula:
C₁₅H₂₇NO₃S MW: 301,4500 g/mol |
Número MDL:
MFCD00011831 Núm. CAS: 733-44-8 EINECS: 211-992-1 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...