(1R,3S)-(+)-Camphoric acid ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: D(+)-Camphoric acid, (1R,3S)-1,2,2-Trimethylcyclopentane-1,3-dicarboxylic acid, Levocarnitine Impurity B

C0012-25G C0012-100G
TCIAC0012-25GEA 49 EUR
TCIAC0012-25G TCIAC0012-100G
(1R,3S)-(+)-Camphoric acid ≥98.0% (by GC, titration analysis)
(1R,3S)-(+)-Camphoric acid
Formula: C₁₀H₁₆O₄
MW: 200,23 g/mol
Número MDL: MFCD00001375
Núm. CAS: 124-83-4
EINECS: 204-715-0
Índice Merck: 14,01733
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6518c89a309c8ecf3c67bd34a58824691324bbe5&vid=6518c89a309c8ecf3c67bd34a58824691324bbe5&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 181.0 to 188.0 deg-C
Specific rotation [a]20/D +43.0 to +49.0 deg (C=5, EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR