5-Methyl-2-(tributylstannyl)thiazole ≥90%
Proveedor: Thermo Scientific
|
Peligro
|
H51531.MD
H51531.03
H51531.MDEA
112
EUR
H51531.MD
H51531.03
5-Methyl-2-(tributylstannyl)thiazole ≥90%
5-Methyl-2-(tributylstannyl)thiazole
Purity: 90+%
CAS No.: 848613-91-2
Molecular Formula: C16H31NSSn
Formula Weight: 388.19
Storage Temperature: 2°C to 8°C
Formula:
C₁₆H₃₁NSSn MW: 388,19 g/mol |
Número MDL:
MFCD09025809 Núm. CAS: 848613-91-2 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...