4-(Benzyloxy)benzonitrile ≥98.0% (por GC)
Proveedor: TCI
|
Atención
|
B1365-25G
B1365-5G
TCIAB1365-25GEA
110
EUR
TCIAB1365-25G
TCIAB1365-5G
4-(Benzyloxy)benzonitrile ≥98.0% (por GC)
4-(Benzyloxy)benzonitrile
Formula:
C₁₄H₁₁NO |
Número MDL:
MFCD00079701 Núm. CAS: 52805-36-4 |
|
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 94.0 to 96.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...