3-Methyl-2-(tributylstannyl)pyridine 96%
Proveedor: Thermo Scientific
|
Peligro
|
H51540.MD
H51540.03
H51540.MDEA
112
EUR
H51540.MD
H51540.03
3-Methyl-2-(tributylstannyl)pyridine 96%
3-Methyl-2-(tributylstannyl)pyridine
Purity: 96%
CAS No.: 259807-97-1
Molecular Formula: C18H33NSn
Formula Weight: 382.16
Storage Temperature: 2°C to 8°C
Formula:
C₁₈H₃₃NSn MW: 382,16 g/mol |
Número MDL:
MFCD07787404 Núm. CAS: 259807-97-1 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...