Capillary columns, Rtx®-624
Proveedor: Restek
40924 40925 10968 10969 10970 10971 10972 10973 10974 10975 10982 40924-6850 10968-6850 10970-101 10970-600 10970-6850 10971-1266850 10971-600 10971-6850 10972-042 10972-6850 10973-6850
568-2451EA 632 EUR
568-2451 568-2452 568-2165 568-2166 568-2167 568-2168 568-2169 568-2170 568-2171 568-2172 568-2173 REST40924-6850 REST10968-6850 REST10970-101 REST10970-600 REST10970-6850 REST10971-1266850 REST10971-600 REST10971-6850 REST10972-042 REST10972-6850 REST10973-6850
Capillary columns, Rtx®-624
Columnas de cromatografía
Application-specific columns for volatile organic pollutants. Recommended in U.S. EPA methods for volatile organic pollutants.
Low to midpolarity phase; Crossbond® 6% cyanopropylphenyl/94% dimethyl polysiloxane
Temperature range: –20 to 240 °C
Suitable replacement for: DB-1301, DB-624, VF-1301 ms, VF-624 ms, CP-1301, ZB-624. Equivalent to USP G43 phase.