Anion exchange HPLC columns, ProPac™ SAX-10 and WAX-10
Proveedor: Thermo Fisher Scientific
063448 063454 063464 088770 088771 088775 063470 063703 063707 054997 054998 054999 055150
555-7287EA 1450.29 EUR
555-7287 555-7288 555-7291 555-7445 555-0258 555-7450 555-7292 555-7302 555-7304 555-7276 555-7277 555-7278 555-7249
Anion exchange HPLC columns, ProPac™ SAX-10 and WAX-10
Columnas de cromatografía
These columns can resolve protein isoforms that differ by a single charged residue. A thin, hydrophilic layer prevents unwanted secondary interactions. The grafted anion exchange surface provides pH based selectivity control and fast mass transfer for high efficiency separation and moderate capacity.
  • High efficiency peaks
  • Higher capacity than other columns based on nonporous particles
  • Ideal for characterisation and quality control assessment of closely-related protein variants
Order Now
ESPECIFICACIONES
Environmentally Preferable
Presión máx. 3000 psi
Rango de pH 2 - 12
Temperature limit 60 °C