Headspace screw caps with 18 mm septa
Proveedor: Agilent
8010-0140 8010-0139 8010-0418 8010-0428
HEWL8010-0140EA 72 EUR
HEWL8010-0140 HEWL8010-0139 HEWL8010-0418 HEWL8010-0428
Headspace screw caps with 18 mm septa
Tapas Tapones para viales de cromatografía
Fits PerkinElmer and other headspace autosamplers.
  • Temperatures resistance up to 300 °C for one hour with minimal degradation
  • Leak proof
  • Compatible with solvents such as DMSO, DMF, Tolulene, and Water