Capillary columns, OPTIMA® 1 MS Accent
Proveedor: MACHEREY-NAGEL
725805.15 725801.25 725801.50 725805.30 725806.30 725805.60 725806.60 725802.30 725807.30 725802.60 725807.60
MANA725805.15EA 545 EUR
MANA725805.15 MANA725801.25 MANA725801.50 MANA725805.30 MANA725806.30 MANA725805.60 MANA725806.60 MANA725802.30 MANA725807.30 MANA725802.60 MANA725807.60
Capillary columns, OPTIMA® 1 MS Accent
Columnas de cromatografía
Nonpolar GC phase with ultra-low bleeding.
  • Ideal for ion trap and quadrupol MS detectors
  • It is widely used for environmental analyses, trace analyses, EPA methods, pesticides, PCB, food and drug analyses
  • Maximum temperature for isothermal operation 340 °C
  • Max. temperature for short isotherms in a temperature program 360 °C
100% dimethylpolysiloxane phase.
USP code: G1/G2/G38.