Ginkgolide B

Proveedor: Cayman Chemical

14636-5 14636-10 14636-25 14636-50
CAYM14636-5EA 47.3 EUR
CAYM14636-5 CAYM14636-10 CAYM14636-25 CAYM14636-50
Ginkgolide B
Ginkgolide B
Formula: C₂₀H₂₄O₁₀
Número MDL: MFCD05662347
Núm. CAS: 15291-77-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR