4-Decyloxybenzoic acid ≥98.0% (by GC, titration analysis)
Proveedor: TCI
D4176-5G
D4176-25G
TCIAD4176-5GEA
66
EUR
TCIAD4176-5G
TCIAD4176-25G
4-Decyloxybenzoic acid ≥98.0% (by GC, titration analysis)
4-Decyloxybenzoic acid
Formula:
C₁₇H₂₆O₃ Pto. de fusión: 96°C |
Número MDL:
MFCD00020360 Núm. CAS: 5519-23-3 |
|
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...