N-Benzyloxycarbonyl-L-alaninol ≥98.0% (by HPLC, total nitrogen)

Proveedor: TCI

Sinónimos: Z-Ala-ol, (S)-Benzyl (1-hydroxypropan-2-yl)carbamate, (S)-(2-Hydroxy-1-methylethyl)carbamic acid benzyl ester, N-Cbz-L-Alaninol

C2629-5G C2629-1G
TCIAC2629-5GEA 55 EUR
TCIAC2629-5G TCIAC2629-1G
N-Benzyloxycarbonyl-L-alaninol ≥98.0% (by HPLC, total nitrogen)
N-Benzyloxycarbonyl-L-alaninol
Formula: C₁₁H₁₅NO₃
MW: 209.25 g/mol
Número MDL: MFCD00672530
Núm. CAS: 66674-16-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=49be62f655a4868d687e9ad5d21fc681e4b05e2f&vid=49be62f655a4868d687e9ad5d21fc681e4b05e2f&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 81.0 to 85.0 deg-C
Specific rotation [a]20/D -4.0 to -7.0 deg (C=2、EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR