1-(o-Tolyl)biguanide ≥97.0% (by titrimetric analysis)
Proveedor: TCI
|
Peligro
|
T0314-25G
T0314-500G
T0314-100G
TCIAT0314-25GEA
29
EUR
TCIAT0314-25G
TCIAT0314-500G
TCIAT0314-100G
1-(o-Tolyl)biguanide ≥97.0% (by titrimetric analysis)
1-(o-Tolyl)biguanide
Formula:
C₉H₁₃N₅ MW: 191,2330 g/mol |
Número MDL:
MFCD00019731 Núm. CAS: 93-69-6 EINECS: 202-268-6 |
|
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 143.0 to 147.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
![](/m-es.vwr.com/es_ES/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...