1,1-Cyclohexanediacetic acid monoamide ≥98.0% (by titrimetric analysis)

Proveedor: TCI

Sinónimos: 2-(1-(2-Amino-2-oxoethyl)cyclohexyl)acetic acid, 2-[1-(2-Amino-2-oxoethyl)cyclohexyl]acetic Acid

C2096-25G C2096-5G
TCIAC2096-25GEA 151 EUR
TCIAC2096-25G TCIAC2096-5G
1,1-Cyclohexanediacetic acid monoamide ≥98.0% (by titrimetric analysis)
1,1-Cyclohexanediacetic acid monoamide
Formula: C₁₀H₁₇NO₃
MW: 199,2500 g/mol
Número MDL: MFCD02181086
Núm. CAS: 99189-60-3
EINECS: 449-430-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9152b978003e8419cb1476862646d6777cdfdc1c&vid=9152b978003e8419cb1476862646d6777cdfdc1c&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 145.0 to 149.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR