2-Chloroethanesulphonic acid sodium salt ∼13% in water

Proveedor: TCI

Sinónimos: Sodium-2-chloroethanesulphonate

C1110-25G C1110-500G
TCIAC1110-25GEA 27 EUR
TCIAC1110-25G TCIAC1110-500G
2-Chloroethanesulphonic acid sodium salt ∼13% in water
2-Chloroethanesulphonic acid sodium salt
Formula: C₂H₄ClNaO₃S
MW: 166,55 g/mol
Número MDL: MFCD00007531
Núm. CAS: 15484-44-3
EINECS: 239-508-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bfcf973792cc0c4b48f18816e03f134d84714b4e&vid=ab8d62ddca36ed021e2297abafc66330ca99c74e&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance Colorless to Light orange to Yellow clear liquid
Concentration (Argentmetric Titration) 13.0 to 15.0 w/w%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR