N-Boc-L-tyrosine ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
Sinónimos:
BOC-L-Tyrosine, Boc-Tyr-OH
B1640-25G
B1640-5G
TCIAB1640-25GEA
113
EUR
TCIAB1640-25G
TCIAB1640-5G
N-Boc-L-tyrosine ≥98.0% (by HPLC, titration analysis)
N-Boc-L-tyrosine
Formula:
C₁₄H₁₉NO₅ MW: 281,3 g/mol |
Número MDL:
MFCD00037179 Núm. CAS: 3978-80-1 EINECS: 223-613-7 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Specific rotation [a]20/D | +1.0 to +4.0 deg (C=2, AcOH) |
Solubility in Acetic acid | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...