2-Fluoro-α-methyl-4-biphenylacetic acid ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Peligro
|
Sinónimos:
Ansaid, 2-(2-Fluoro-[1,1'-biphenyl]-4-yl)propanoic acid, Flurbiprofen, Froben, Antadys
F0371-25G
F0371-5G
TCIAF0371-25GEA
240
EUR
TCIAF0371-25G
TCIAF0371-5G
2-Fluoro-α-methyl-4-biphenylacetic acid ≥98.0% (by HPLC, titration analysis)
2-Fluoro-α-methyl-4-biphenylacetic acid
Formula:
C₁₅H₁₃FO₂ MW: 244.265 g/mol |
Número MDL:
MFCD00079303 Núm. CAS: 5104-49-4 EINECS: 225-827-6 UN: 2811 ADR: 6.1 III Índice Merck: 14,04199 |
|
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 113.0 to 116.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...