3-(tert-Butyldimethylsilyloxy)phenylboronic acid (contains varying amounts of Anhydride)
Proveedor: TCI
|
Atención
|
Sinónimos:
3-(tert-Butyl dimethylsiloxy)phenyl boronic acid, (3-((tert-Butyldimethylsilyl)oxy)phenyl)boronic acid, 3-(tert-Butyldimethylsiloxy)benzeneboronic acid
B3726-1G
B3726-5G
TCIAB3726-1GEA
61
EUR
TCIAB3726-1G
TCIAB3726-5G
3-(tert-Butyldimethylsilyloxy)phenylboronic acid (contains varying amounts of Anhydride)
3-(tert-Butyldimethylsilyloxy)phenylboronic acid
Formula:
C₁₂H₂₁BO₃Si MW: 252.19 g/mol |
Número MDL:
MFCD03701505 Núm. CAS: 261621-12-9 |
|
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 108.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...