1,3-Bis(3-aminophenoxy)benzene ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

B1370-25G B1370-5G
TCIAB1370-25GEA 208 EUR
TCIAB1370-25G TCIAB1370-5G
1,3-Bis(3-aminophenoxy)benzene ≥98.0% (by HPLC, titration analysis)
1,3-Bis(3-aminophenoxy)benzene
Formula: C₁₈H₁₆N₂O₂
MW: 292,3318 g/mol
Número MDL: MFCD00043718
Núm. CAS: 10526-07-5
EINECS: 234-082-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=37d78a4da8a37a9166cb78f811cf0af2dc6b48ac&vid=37d78a4da8a37a9166cb78f811cf0af2dc6b48ac&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 107.0 to 109.0 deg-C
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR