4-Cyanophenyl-4-heptylbenzoate ≥98.0% (por GC)
Proveedor: TCI
H0810-1G
H0810-5G
TCIAH0810-1GEA
30
EUR
TCIAH0810-1G
TCIAH0810-5G
4-Cyanophenyl-4-heptylbenzoate ≥98.0% (por GC)
4-Cyanophenyl-4-heptylbenzoate
Formula:
C₂₁H₂₃NO₂ |
Número MDL:
MFCD00001817 Núm. CAS: 38690-76-5 EINECS: 254-084-0 |
|
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Mesomorphic range | 42.0 to 61.0 deg-C |
Solubility in Toluene | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...