4-Methyl-N,N-diphenylaniline ≥98.0% (por GC)
Proveedor: TCI
|
Atención
|
M1898-1G
M1898-5G
TCIAM1898-1GEA
56
EUR
TCIAM1898-1G
TCIAM1898-5G
4-Methyl-N,N-diphenylaniline ≥98.0% (por GC)
4-Methyl-N,N-diphenylaniline
Formula:
C₁₉H₁₇N MW: 259.3450 g/mol |
Número MDL:
MFCD03093256 Núm. CAS: 4316-53-4 |
|
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 66.0 to 72.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...