Potassium(3-pyridyl)trifluoroborate ≥98.0% (by total nitrogen basis)
Proveedor: TCI
|
Atención
|
Sinónimos:
Potassium trifluoro(pyridin-3-yl)borate
P1717-1G
P1717-5G
TCIAP1717-1GEA
43
EUR
TCIAP1717-1G
TCIAP1717-5G
Potassium(3-pyridyl)trifluoroborate ≥98.0% (by total nitrogen basis)
Potassium(3-pyridyl)trifluoroborate
Formula:
C₅H₄BF₃KN MW: 185,0000 g/mol |
Número MDL:
MFCD09038545 Núm. CAS: 561328-69-6 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...