Tantalum tris(diethylamido)-tert-butylimide ≥99.99% (metals basis)

Proveedor: Thermo Fisher Scientific
Peligro


Ampuled under argon

43419.14 43419.06
43419.14EA 976 EUR
43419.14 43419.06
Tantalum tris(diethylamido)-tert-butylimide ≥99.99% (metals basis)
Tantalum tris(diethylamido)-tert-butylimide

Purity: 99.99% CAS No.: 169896-41-7 Molecular Formula: C16H39N4Ta Formula Weight: 468.48 Physical Form: Liquid, Ampouled under argon Sensitivity: Air and Moisture Sensitive Appearance: Yellow Boiling Point: 90°/0.1mm UN No.: 3129 MDL No.: MFCD02684506


Precursor for TaN

Formula: C₁₆H₃₉N₄Ta
Pto. Ebullición: 90 °C (0,1 mmHg)
Temperatura de almacenaje: Ambiente
Número MDL: MFCD02684506
Núm. CAS: 169896-41-7
UN: 3129
ADR: 4.3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR