Picolinic acid hydrochloride ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: Pyridine-2-carboxylic acid hydrochloride, Picolinic acid hydrochloride

P0422-25G P0422-5G
TCIAP0422-25GEA 125 EUR
TCIAP0422-25G TCIAP0422-5G
Picolinic acid hydrochloride ≥98.0% (by HPLC, titration analysis)
Picolinic acid hydrochloride
Formula: C₆H₆ClNO₂
MW: 159,5703 g/mol
Número MDL: MFCD00050666
Núm. CAS: 636-80-6
EINECS: 211-266-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e7d3b1e0a8594af63f78117094d88e232c5cb55&vid=1e7d3b1e0a8594af63f78117094d88e232c5cb55&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR