6-Methylpicolinic acid ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 6-Methyl-2-pyridinecarboxylic acid, 6-Methylpyridine-2-carboxylic acid

M0873-25G M0873-5G
TCIAM0873-25GEA 126 EUR
TCIAM0873-25G TCIAM0873-5G
6-Methylpicolinic acid ≥98.0% (by HPLC, titration analysis)
6-Methylpicolinic acid
Formula: C₇H₇NO₂
MW: 137,14 g/mol
Número MDL: MFCD00023481
Núm. CAS: 934-60-1
EINECS: 213-287-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2a292e51a0143b3fe9b6925920ce6900813ffed0&vid=2a292e51a0143b3fe9b6925920ce6900813ffed0&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 127.0 to 132.0 deg-C
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR