6-Amino-2-methylquinoline ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
2-methylquinolin-6-amine
A1047-1G
A1047-5G
TCIAA1047-1GEA
154
EUR
TCIAA1047-1G
TCIAA1047-5G
6-Amino-2-methylquinoline ≥98.0% (by GC, titration analysis)
6-Amino-2-methylquinoline
Formula:
C₁₀H₁₀N₂ MW: 158,20 g/mol |
Número MDL:
MFCD00052600 Núm. CAS: 65079-19-8 |
Resultados test especificaciones
Appearance | Light yellow to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 187.0 to 192.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...