Cesium wolframate ≥99.9% (metals basis)
Proveedor: Thermo Scientific
Sinónimos:
Cesium tungsten oxide, Cesium tungstate
39206.06
39206.14
39206.06EA
98
EUR
39206.06
39206.14
Cesium wolframate ≥99.9% (metals basis)
Cesium wolframate
Purity: 99.9%
CAS No.: 13587-19-4
Molecular Formula: Cs2O4W
Formula Weight: 513.66
Physical Form: -200 Mesh Powder
Sensitivity: Hygroscopic
Appearance: White to off-white
Odour: Odourless
MDL No.: MFCD00049438
Formula:
Cs₂O₄W MW: 513,66 g/mol Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00049438 Núm. CAS: 13587-19-4 EINECS: 237-019-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...