Potassium wolframate ≥99.5% (metals basis)
Proveedor: Thermo Fisher Scientific
Sinónimos:
Potassium tungstate, Potassium tungsten oxide
14031.14
14031.22
14031.14EA
86.5
EUR
14031.14
14031.22
Potassium wolframate ≥99.5% (metals basis)
Potassium wolframate
Purity: 99.5%
CAS No.: 7790-60-5
Molecular Formula: K2WO4
Formula Weight: 326.04
Physical Form: -60 Mesh Powder
Density: 3.12
Melting Point: 921°
Solubility: Soluble in water
Merck Reference: 14,7694
MDL No.: MFCD00049668
Formula:
K₂O₄W MW: 326.03 g/mol Pto. de fusión: 921 °C Densidad: 3.12 Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00049668 Núm. CAS: 7790-60-5 EINECS: 232-215-2 Índice Merck: 14,7694 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...