5,5'-Dimethyl-2,2'-bipyridyl ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
5,5'-Dimethyl-2,2'-bipyridine, Abametapir, 6,6'-Bi-3-picoline
B2138-1G
B2138-25G
B2138-5G
TCIAB2138-1GEA
30
EUR
TCIAB2138-1G
TCIAB2138-25G
TCIAB2138-5G
5,5'-Dimethyl-2,2'-bipyridyl ≥98.0% (by GC, titration analysis)
5,5'-Dimethyl-2,2'-bipyridyl
Formula:
C₁₂H₁₂N₂ MW: 184,24 g/mol |
Número MDL:
MFCD01740554 Núm. CAS: 1762-34-1 |
Resultados test especificaciones
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 113.0 to 117.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...