2-Thiazolecarboxylic acid 95%
Proveedor: Thermo Fisher Scientific
|
Atención
|
Sinónimos:
Thiazole-2-carboxylic acid
H63620.03
H63620.06
H63620.03EA
95.6
EUR
H63620.03
H63620.06
2-Thiazolecarboxylic acid 95%
2-Thiazolecarboxylic acid
Purity: 95%
CAS No.: 14190-59-1
Molecular Formula: C4H3NO2S
Formula Weight: 129.14
Storage Temperature: -30°C to -10°C
Physical Form: Solid
MDL No.: MFCD02094169
Formula:
C₄H₃NO₂S MW: 129.14 g/mol |
Número MDL:
MFCD02094169 Núm. CAS: 14190-59-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...