Capillary columns, Rxi®-5Sil MS (fused silica)
Proveedor: Restek
43602 13650 13624 13623 13626 13624-6850 43604 13620 43601 43605 43607 43815 43816 43817
REST43602EA 744 EUR
REST43602 REST13650 REST13624 REST13623 REST13626 REST13624-6850 REST43604 REST13620 REST43601 REST43605 REST43607 REST43815 REST43816 REST43817
Capillary columns, Rxi®-5Sil MS (fused silica)
Columnas de cromatografía
Rxi®-5Sil MS stationary phase incorporates phenyl groups in the polymer backbone. This improves thermal stability, reduces bleed, and makes the phase less prone to oxidation. These columns are ideal for GC-MS applications requiring high sensitivity, including use in ion trap systems.
  • Low-bleed GC-MS column
  • Low polarity phase; Crossbond 1,4-bis(dimethylsiloxy)phenylene dimethyl polysiloxane columns
  • Temperature range: −60 to 350 °C
Ideal for semivolatiles, polycyclic aromatic compounds, chlorinated hydrocarbons, phthalates, phenols, amines, organochlorine pesticides, organophosphorus pesticides, drugs, solvent impurities, and hydrocarbons.