Chemical resistant overalls, Spacel 3000 AP/RA/EBJ, PE
Proveedor: HONEYWELL SAFETY
4503003-M 4503003-S 4503003-XL 4503003-XXL 4503003-XXXL
DALL4503003-MEA 0 EUR
DALL4503003-M DALL4503003-S DALL4503003-XL DALL4503003-XXL DALL4503003-XXXL
Chemical resistant overalls, Spacel 3000 AP/RA/EBJ, PE
Prendas desechables
Liquid tight disposable coverall designed to protect workers from chemicals. Made of three-layered yellow polyethylene film.
  • New ergonomics offer a better fitting and protection
  • Elasticated wrists and ankles, double adhesive flap and back ventilation valve
  • Very good abrasion, tear and puncture resistance
  • Watertight due to 'Spaseal' welding
  • Yellow colour for improved visibility
Ideal for use in chemical industries, petrochemistry, industrial maintenance, pharmaceutical industry, spoiled beach cleaning, transportation of hazardous materials, industrial sub-contracting cleaning, sanitation, waste processing and emergency services.
Thickness: 100 µm
Certificaciones: Type 4 + 6; Cat. III.