Cell culture media, Glasgow MEM (GMEM BHK 21)
Proveedor: Biowest
P0120-N1L P0120-N10L P0120-N50L
P0120-N1LEA 26.6 EUR
P0120-N1L P0120-N10L P0120-N50L
Cell culture media, Glasgow MEM (GMEM BHK 21)
Medios de cultivo celular
Glasgow MEM (GMEM) was originally formulated for growing BHK 21 cells. The original formulation is an Eagle's Basal Medium modified to contain two times the normal concentration of amino acids and vitamins. It is intended for use with adherent cell lines.