Cut resistant gloves, Tuff Cut Nit
Proveedor: HONEYWELL SAFETY
2232533/10 2232533/8 2232533/9
DALL2232533/10EA 0 EUR
DALL2232533/10 DALL2232533/8 DALL2232533/9
Cut resistant gloves, Tuff Cut Nit
Guantes
These gloves are knitted from Kevlar® yarn combined with stainless steel fibre. They feature a black nitrile foam coating on the palm.
  • Stainless steel fibres provide maximum protection
  • Long sleeve for increased protection of the wrist
  • Nitrile coating provides good protection against oil and grease
  • Foam coating is more flexible than standard nitrile coating
Ideal for handling heavy, very sharp parts with cutting edges, e.g. paper, metal and glass sheets and plastic parts. Also suitable for finishing operations with cutters.
Certificaciones: EN 388:20More Product Information
Order Now
ESPECIFICACIONES
Modelo Nit
Environmentally Preferable
Ambidiestro/Específico para cada mano Específico para cada mano
Desechable / Reutilizable Reutilizable
Material del revestimiento Kevlar®
Acabado Coating on palm and fingers
Calibre 10
Tipo de puño Puño elástico
Normativa EN EN 388:2016 (4X43D), EN 407 (X1XXXX)