Laboratory/media bottles, wide neck, with or without screw cap, BIBBY®
Proveedor: SciLabware
1523/06 1523/08 1523/10 1523/12 1523/14 1523/16 1523/18 1523/22
BIBB1523/06EA 166 EUR
BIBB1523/06 BIBB1523/08 BIBB1523/10 BIBB1523/12 BIBB1523/14 BIBB1523/16 BIBB1523/18 BIBB1523/22
Laboratory/media bottles, wide neck, with or without screw cap, BIBBY®
Frascos Botellas de vidrio
PYREX®, Borosilicate glass, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Available without screw cap or with screw cap and pouring ring.