Chemical resistant gloves, NitoPren® 717
Proveedor: KCL by Honeywell
717.10 717.8 717.9 717.7 717.11
KCHL717.10EA 11.8 EUR
KCHL717.10 KCHL717.8 KCHL717.9 KCHL717.7 KCHL717.11
Chemical resistant gloves, NitoPren® 717
Guantes
These protective gloves are made of polychloroprene and nitrile. The two-layer system offers a high resistance against a variety of different groups of hazardous substances.
  • Good grip when handling wet and oily parts
  • Free from natural latex
  • Anatomical fit allows working without user fatigue
  • Flock lining reduces sweating
Ideal for use in the chemical and pharmaceutical industry, in metal working and finishing, in the printing industry and in laboratories.
Certificaciones: EN 388:2016 (3001X), EN ISO 374-1 Type A (OTPAKL), EN ISO 374-5 VIRUS; Cat. III; AQL 0,65
Order Now
ESPECIFICACIONES
Modelo 717
Environmentally Preferable
Ambidiestro/Específico para cada mano Específico para cada mano
Desechable / Reutilizable Reutilizable
Latex/No contiene Latex No contiene Latex
Acabado Smooth
Tipo de puño Puño con reborde
Normativa EN EN 388:2016 (3001X), EN ISO 374-1 Type A (OTPAKL), EN ISO 374-5 VIRUS