EasyLOK fittings
Proveedor: Trajan Scientific and Medical
206102 206160
SGEA206102EA 53 EUR
SGEA206102 SGEA206160
EasyLOK fittings
Racores y conectores
These fittings are composed of a knurled stainless steel nut and a double ended PEEK™ ferrule.
The PEEK™ ferrule simply slides over any 1,58 mm OD tubing to its required position, while the nut is finger tightened. It will not crush the tubing and can be easily readjusted for quick column changes. The unique double ended ferrule design seals at two points to prevent leaks.
The fittings are compatible with any standard female HPLC fitting, including Swagelok®, Parker™, Waters®, Valco® and Whatman®. Also used with SGE GLT™ columns.