Capillary columns, MolSieve 13X
Proveedor: Agilent
G3591-70017 G3591-70015 G3591-81028 G3591-81031 G3591-81043 G3591-81054 G3591-81085 G3591-81097 G3591-81098 G3591-81101 G3591-82003 G3591-82012 G3591-82043 G3591-82097 G3591-82101 G3591-80097 G3591-81058 G3591-80003 G3591-80012 G3591-80028 G3591-80031 G3591-80043 G3591-80054 G3591-80058 G3591-80085 G3591-80098 G3591-80101 G3591-81003 G3591-81012
HEWLG3591-70017EA 369 EUR
HEWLG3591-70017 HEWLG3591-70015 HEWLG3591-81028 HEWLG3591-81031 HEWLG3591-81043 HEWLG3591-81054 HEWLG3591-81085 HEWLG3591-81097 HEWLG3591-81098 HEWLG3591-81101 HEWLG3591-82003 HEWLG3591-82012 HEWLG3591-82043 HEWLG3591-82097 HEWLG3591-82101 HEWLG3591-80097 HEWLG3591-81058 HEWLG3591-80003 HEWLG3591-80012 HEWLG3591-80028 HEWLG3591-80031 HEWLG3591-80043 HEWLG3591-80054 HEWLG3591-80058 HEWLG3591-80085 HEWLG3591-80098 HEWLG3591-80101 HEWLG3591-81003 HEWLG3591-81012
Capillary columns, MolSieve 13X
Columnas de cromatografía
These MolSieve 13X columns have wide range of tubing materials including stainless steel, ultimetal and nickel plus hundreds of stationary phases, packings, and supports to suit many specific application.
  • UltiMetal treated stainless steel tubing allows for improved inertness and peak shape performance
  • Rigorous, highly efficient packing technology ensures column-to-column reproducibility