Colorimeters, monoparameter, handheld, Checker® HC series
Proveedor: Hanna
Hanna pocket checkers offer a cost-effective digital alternative to visual chemical test kits. Ideal for regular water testing, swimming pool testing and for aquaculture hobbyists.
- Testing is quick and easy
- High precision results – more accurate than visual chemical test kits
- Auto shut off feature means the Checker cannot be left on by mistake
- Fully portable, fitting easily into both palm and pocket
Información de suministro: Colorimeters supplied with two sample cuvettes, starter pack of reagents, 1×1,5 V AAA battery and instruction manual.
Precaución: *PCU= Platinum cobalt units