Occupational shoes, Velcro® fastening, Naturform, Kempten
Proveedor: SCHURR SCHUHVERTRIEB
11433.01.35/38 11433.01.35/39 11433.01.35/40 11433.01.35/37
SCRR11433.01.35/38EA 0 EUR
SCRR11433.01.35/38 SCRR11433.01.35/39 SCRR11433.01.35/40 SCRR11433.01.35/37
Occupational shoes, Velcro® fastening, Naturform, Kempten
Zapatos y botas
Occupational shoe made from full grained leather upper, leather lining, padded ankle collar and a rubber PU sole with anti-slip resistance.
  • Exchangeable insole made from PUR foam, orthopaedic insole possible
  • Anti-slip and anti static A4 sole
  • Triple plus shock absorption in sole
Ideal for use in kitchens and for food processing.
Colour: White
Certificaciones: EN ISO 20347:2004; O1