Manual crimpers and decappers
Proveedor: Agilent
5181-8827 8710-1643 5040-4667 5040-4668 5040-4669 5040-4671 5190-4062 5190-4064 5190-4065
HEWL5181-8827EA 457 EUR
HEWL5181-8827 HEWL8710-1643 HEWL5040-4667 HEWL5040-4668 HEWL5040-4669 558-0571 HEWL5190-4062 HEWL5190-4064 HEWL5190-4065
Manual crimpers and decappers
Encapsuladores y desencapsuladores
Crimpers and decappers for 8, 11 and 20 mm crimp top vials and crimp caps.