Thermostatic baths for viscometers, CT 72
Proveedor: SI Analytics
285418526 285418547 285418568 285418501 285418615
SCOI285418526EA 6510 EUR
SCOI285418526 SCOI285418547 645-0081 645-0080 SCOI285418615
Thermostatic baths for viscometers, CT 72
Baños Baños de viscosímetro
The CT 72 thermostatic baths are designed for determining viscosity of newtonian liquids in glass capillary viscometers with the display of the momentary and the set temperature through VF display.
  • User friendly through automatic fuses on the back panel instead of micro fuses
  • Programmable set temperatures through integrated clock with program controller
  • Easy to reach overtemperature safety system on front panel
Order Now
ESPECIFICACIONES
Modelo CT 72/P CT 72/2 CT 72/4 CT 72/E CT 72/2-TT
Environmentally Preferable
Capacidad 15 L 15 L 27 L 15 L 15 L
Intervalo de temperaturas 10...60 °C 5...150 °C 5...150 °C 10...60 °C –40...+150 °C
Posiciones del viscómetro 2 2 4 2 2
Material del baño Stainless steel Stainless steel Stainless steel and glass Stainless steel Stainless steel
W×D×H 355×370×250 mm 355×370×250 mm 605×250×370 mm 355×370×250 mm 355×370×250 mm
Peso 5 kg 13,5 kg 28 kg 5 kg 14 kg