Bottles, narrow neck, round, PC, Boston type, with screw caps, Azlon®
Proveedor: SciLabware
BRC0500 BRC1000
215-3597EA 270 EUR
215-3597 215-3598
Bottles, narrow neck, round, PC, Boston type, with screw caps, Azlon®
Frascos Botellas de plástico
Polycarbonate, highly transparent.
  • Leakproof
  • Suitable for repeated sterilisation by autoclaving at +121 °C
  • Heavy-duty and durable
Ideal for the preparation and storage of biological media, reagents and aqueous solutions. With linerless polypropylene cap.